Thermo Scientific Chemicals

Potassium heptafluorotantalate(V), 99.7%, Thermo Scientific Chemicals

Catalog number: 040316.30
250 g, Each
Thermo Scientific Chemicals

Potassium heptafluorotantalate(V), 99.7%, Thermo Scientific Chemicals

Catalog number: 040316.30
250 g, Each
Quantity
Catalog number: 040316.30
also known as 040316-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16924-00-8
IUPAC Name
dipotassium heptafluorotantalumdiuide
Molecular Formula
F7H7K2Ta
InChI Key
BLXHHPZXEPALDX-UHFFFAOYSA-N
SMILES
F.F.F.F.F.F.F.[K+].[K+].[Ta--]
Assay (unspecified)
99.7% min

Description

It is an important raw material. Used in chemical research. Use as primary and secondary intermediate(Pharmaceutical Intermediates). Used in the production and preparation of tantalum powder.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text