Thermo Scientific Chemicals

Cobalt(II) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040319.06
5 g, Each
Thermo Scientific Chemicals

Cobalt(II) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040319.06
5 g, Each
Quantity
Catalog number: 040319.06
also known as 040319-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-43-7
IUPAC Name
λ²-cobalt(2+) dibromide
Molecular Formula
Br2Co
InChI Key
BZRRQSJJPUGBAA-UHFFFAOYSA-L
SMILES
[Co++].[Br-].[Br-]
Residual water
< 100 ppm
Particle Size
-10 Mesh
Appearance (Color)
Green
Form
Beads
Total Metal Impurities
0.01% max.

Description

Used primarily as a catalyst in some processes. Chiefly used in hygrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text