Thermo Scientific Chemicals

Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040320.36
500 g, Each
Thermo Scientific Chemicals

Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040320.36
500 g, Each
Quantity
Catalog number: 040320.36
also known as 040320-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-13-3
IUPAC Name
ammonium sodium tetrahydrate hydrogen phosphate
Molecular Formula
H13NNaO8P
InChI Key
IQTQISLCLWJRPM-UHFFFAOYSA-M
SMILES
[NH4+].O.O.O.O.[Na+].OP([O-])([O-])=O
Assay (unspecified)
99% NaNH₄HPO₄?4H₂O min

Description

Used in the laboratory as an essential ingredient of the microcosmic salt bead test for identification of metallic radicals. Also used as intermediates. Finds application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text