Potassium fluorosulfate, 99.5%, Thermo Scientific Chemicals
Potassium fluorosulfate, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium fluorosulfate, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 040322.09
also known as 040322-09
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
Add to cart
Potassium fluorosulfate, 99.5%, Thermo Scientific Chemicals
Catalog number040322.09
Price (USD)/ Each
135.65
Online exclusive
150.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13455-22-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99.5% min
Use as primary and secondary intermediate. Finds application in chemical research..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only