Thermo Scientific Chemicals

Lanthanum boride, 99.5% (REO), Thermo Scientific Chemicals

Catalog number: 040325.30
250 g, Each
Thermo Scientific Chemicals

Lanthanum boride, 99.5% (REO), Thermo Scientific Chemicals

Catalog number: 040325.30
250 g, Each
Quantity
Catalog number: 040325.30
also known as 040325-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12008-21-8
IUPAC Name
pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide lanthanum
Molecular Formula
B6La
InChI Key
IBDHMAYMLSYGAK-UHFFFAOYSA-N
SMILES
[La].B12B3[B-]11B4B1[B-]234
Total Rare Earth Oxide Impurities
0.5% max.

Description

Lanthanum boride is employed in hot cathodes, which enact and includes electron microscopes, microwave tubes, electron lithography, electron beam welding, X-ray tubes, and free electron lasers. It is administered as a size or strain standard in X-ray powder diffraction to calibrate instrumental broadening of diffraction peaks. It exercises as a superconductor. It has a great potential as a thermionic emitter in renewable energy applications. It is widely used in teleran, electronic industry, instrument, medical equipment, metallurgy and environmental protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text