Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals
Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
25 cm
Catalog number 040329.BS
also known as 040329-BS
Price (USD)/ Each
456.00
-
Add to cart
Quantity:
10 cm
Request bulk or custom format
Price (USD)/ Each
456.00
Add to cart
Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals
Catalog number040329.BS
Price (USD)/ Each
456.00
-
Add to cart
Specifications
Chemical Name or MaterialGold wire (14kt)
ColorYellow
Name Note1.63mm (0.064 in.) dia.
Recommended StorageAmbient temperatures
TSCAYes
View more
Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.