Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals

Catalog number: 040331.14
25 g, Each
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals

Catalog number: 040331.14
25 g, Each
Quantity
Catalog number: 040331.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-98-6
Molecular Formula
((C2H5N)zC2H4N)y(C2H5N)x
SMILES
*-CCNCCN(-*)CCN-*
MDL Number
MFCD00084427
Appearance (Color)
Yellow
Viscosity
40,000 - 150,000 cps
Molecular Weight
10,000
Form
Liquid

Description

Polyethyleneimine, branched, M.W. 10,000 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics. It is investigated as non-viral vector carriers in research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text