Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals
Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 040331.36
also known as 040331-36
Price (USD)/ Each
648.65
Online exclusive
720.00 
Save 71.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
648.65
Online exclusive
720.00 
Save 71.35 (10%)
Add to cart
Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals
Catalog number040331.36
Price (USD)/ Each
648.65
Online exclusive
720.00 
Save 71.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Viscosity40,000 - 150,000 cps
Molecular Weight10,000
FormLiquid
Polyethyleneimine, branched, M.W. 10,000 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics. It is investigated as non-viral vector carriers in research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only