Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%, Thermo Scientific Chemicals
Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%, Thermo Scientific Chemicals

Conductive adhesives
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040340.14
also known as 040340-14
Price (USD)/ Each
56.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
66.80 
Save 10.15 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
56.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
66.80 
Save 10.15 (15%)
Add to cart
Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%, Thermo Scientific Chemicals
Catalog number040340.14
Price (USD)/ Each
56.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
66.80 
Save 10.15 (15%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAluminum
Name NoteSilver coated powder
CAS11145-27-0
Recommended StorageAmbient temperatures
TSCAYes
View more
Conductive adhesives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only