Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals
Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040342.09
also known as 040342-09
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.10 
Save 10.45 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.10 
Save 10.45 (15%)
Add to cart
Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals
Catalog number040342.09
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
69.10 
Save 10.45 (15%)
-
Add to cart
Chemical Identifiers
CAS7446-18-6
IUPAC Namebis(λ¹-thallanylium) sulfate
Molecular FormulaO4STl2
InChI KeyYTQVHRVITVLIRD-UHFFFAOYSA-L
SMILES[Tl+].[Tl+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only