Chemical Name or MaterialSulfur in Heavy Mineral Oil standard solution
Name Note500μg/g (0.0500%)
Health Hazard 1H304
Quantity100 mL
Health Hazard 2GHS H Statement H350 May cause cancer.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.