Thermo Scientific Chemicals
Sulfur in Heavy Mineral Oil standard solution, Specpure™, 750μg/g (0.0750%), Thermo Scientific Chemicals
Catalog number: 040350.AE
100 mL, Each
Thermo Scientific Chemicals
Sulfur in Heavy Mineral Oil standard solution, Specpure™, 750μg/g (0.0750%), Thermo Scientific Chemicals
Catalog number: 040350.AE
100 mL, Each
Quantity
Catalog number: 040350.AE
also known as 040350-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
750?g/g ?1% (0.0750 wt%)
Starting Material
di-n-butyl sulfide (nDBS)