2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Thermo Scientific Chemicals
2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 040366.36
also known as 040366-36
Price (USD)/ Each
33.65
Online exclusive
37.20 
Save 3.55 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
37.20 
Save 3.55 (10%)
Add to cart
2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Thermo Scientific Chemicals
Catalog number040366.36
Price (USD)/ Each
33.65
Online exclusive
37.20 
Save 3.55 (10%)
-
Add to cart
Chemical Identifiers
CAS25265-77-4
IUPAC Name3-hydroxy-2,2,4-trimethylpentyl 2-methylpropanoate
Molecular FormulaC12H24O3
InChI KeyDAFHKNAQFPVRKR-UHFFFAOYNA-N
SMILESCC(C)C(O)C(C)(C)COC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
FormLiquid
Refractive Index1.4411 to 1.4433
Specific Gravity0.9450 to 0.9550
Assay (unspecified)> 98.5 wt% ester alcohol
Employed In pharma industry. As Coalescent for latex paints. Finds applications in Coatings, Nail care, Printing inks, Solvents for cosmetics and personal care, plasticizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only