Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 040367.14
also known as 040367-14
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
Add to cart
Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number040367.14
Price (USD)/ Each
216.65
Online exclusive
240.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12036-22-5
IUPAC Namedioxotungsten
Molecular FormulaO2W
InChI KeyDZKDPOPGYFUOGI-UHFFFAOYSA-N
SMILESO=[W]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Tungsten(IV) oxide is used in the production of tungsten and tungstates, in alloys, in fire-proofing fabrics and as a ceramic pigment. It is used to study their chromism, photocatalysis and sensing capabilities. It is associated with rhenium trioxide particles and used for detection of protonatable, radical and precharged species under laser desorption ionization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only