Thermo Scientific Chemicals

Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040367.22
100 g, Each
Thermo Scientific Chemicals

Tungsten(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040367.22
100 g, Each
Quantity
Catalog number: 040367.22
also known as 040367-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12036-22-5
IUPAC Name
dioxotungsten
Molecular Formula
O2W
InChI Key
DZKDPOPGYFUOGI-UHFFFAOYSA-N
SMILES
O=[W]=O
Total Metal Impurities
0.1% max.

Description

Tungsten(IV) oxide is used in the production of tungsten and tungstates, in alloys, in fire-proofing fabrics and as a ceramic pigment. It is used to study their chromism, photocatalysis and sensing capabilities. It is associated with rhenium trioxide particles and used for detection of protonatable, radical and precharged species under laser desorption ionization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text