Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number 040373.14
also known as 040373-14
Price (USD)/ Each
868.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
868.00
Add to cart
Cerium(III) bromide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals
Catalog number040373.14
Price (USD)/ Each
868.00
-
Add to cart
Chemical Identifiers
CAS14457-87-5
IUPAC Namecerium(3+) tribromide
Molecular FormulaBr3Ce
InChI KeyMOOUSOJAOQPDEH-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total Rare Earth Oxide Impurities0.1% max.
Residual water< 100 ppm
Particle Size-10 Mesh
FormBeads
Cerium(III) bromide is used as an additive for chemical reagent. Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only