Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase, Thermo Scientific Chemicals
Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number 040379.30
also known as 040379-30
Price (USD)/ Each
478.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
478.00
Add to cart
Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase, Thermo Scientific Chemicals
Catalog number040379.30
Price (USD)/ Each
478.00
-
Add to cart
Chemical Identifiers
CAS12033-89-5
IUPAC NameSilicon nitride
Molecular FormulaN4Si3
InChI KeyHQVNEWCFYHHQES-UHFFFAOYSA-N
SMILESN12[Si]34N5[Si]11N3[Si]25N41
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.7% max., typically 90% ?-phase
Silicon(IV) nitride is used in automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only