Thermo Scientific Chemicals
Boron nitride, Aerosol Refractory Paint, 97+%, (balance B{2}O{3}), Thermo Scientific Chemicals
Catalog number: 040389.0CL
5 x 13 oz., Each
Thermo Scientific Chemicals
Boron nitride, Aerosol Refractory Paint, 97+%, (balance B{2}O{3}), Thermo Scientific Chemicals
Catalog number: 040389.0CL
5 x 13 oz., Each
Quantity
Catalog number: 040389.0CL
also known as 040389-0:
Price (USD)
Quantity
-
Have Questions?
Description
Boron nitride is used to make alloys, semiconductors, nuclear reactors and lubricants. It is also used in the fabrication and luminescence study of monolayered boron nitride quantum dots. Further, it acts as a starting material to form boron nitride nanotubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use