Thermo Scientific Chemicals

Bismuth subsalicylate, 96%, Thermo Scientific Chemicals

Catalog number: 040395.30
250 g, Each
Thermo Scientific Chemicals

Bismuth subsalicylate, 96%, Thermo Scientific Chemicals

Catalog number: 040395.30
250 g, Each
Quantity
Catalog number: 040395.30
also known as 040395-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14882-18-9
IUPAC Name
4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
Molecular Formula
C7H6BiO4
InChI Key
QBWLKDFBINPHFT-UHFFFAOYSA-L
SMILES
O.O=C1O[Bi]OC2=CC=CC=C12
Appearance (Color)
White to off-white
Form
Powder
Assay (unspecified)
≥54.8% bismuth dried basis (theory: ≥95% product)

Description

To impart pearly surface to cellulose-base, polystyrene, and phenolformaldehyde resins. Typical tap bulk density 0.43g/cm^3Bismuth subsalicylate is used to treat temporary discomforts of the stomach and gastrointestinal tract, such as diarrhea, indigestion, heartburn and nausea. It also acts as an antacid. It is practiced to decrease the number of bowel movements and to make the stool firmer. It is also used to reduce inflammation within the intestine. It is further used in growth media for selective isolation of microorganisms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text