Thermo Scientific Chemicals

Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals

Catalog number: 040396.18
50 g, Each
Thermo Scientific Chemicals

Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals

Catalog number: 040396.18
50 g, Each
Quantity
Catalog number: 040396.18
also known as 040396-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-59-9
IUPAC Name
chlorobismuthanone
Molecular Formula
BiClH2O
InChI Key
OMRKLWMUNWPRFB-UHFFFAOYSA-N
SMILES
Cl.O=[BiH]
Loss on Drying
1% max

Description

Bismuth(III) chloride oxide is used in pharmaceutical industry, in the production of cosmetics, in chemical and metallurgical industries. It is used as pigment, in manufacture of artificial pearls and as a dry-cell cathodes, in photocatalysts for degradation of organic pollutant, in white-light LEDs, in the manufacture of plastics, in the synthesis of methanol, in semiconductor devices such as CCDs or photodiodes and also used in the production of permanent magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text