Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals
Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals

Used as pigment, in manufacture of artificial pearls, dry-cell cathodes
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Catalog number 040396.30
also known as 040396-30
Price (USD)/ Each
178.65
Online exclusive
198.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
178.65
Online exclusive
198.00 
Save 19.35 (10%)
Add to cart
Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals
Catalog number040396.30
Price (USD)/ Each
178.65
Online exclusive
198.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7787-59-9
IUPAC Namechlorobismuthanone
Molecular FormulaBiClH2O
InChI KeyOMRKLWMUNWPRFB-UHFFFAOYSA-N
SMILESCl.O=[BiH]
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying1% max
Bismuth(III) chloride oxide is used in pharmaceutical industry, in the production of cosmetics, in chemical and metallurgical industries. It is used as pigment, in manufacture of artificial pearls and as a dry-cell cathodes, in photocatalysts for degradation of organic pollutant, in white-light LEDs, in the manufacture of plastics, in the synthesis of methanol, in semiconductor devices such as CCDs or photodiodes and also used in the production of permanent magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only