Thermo Scientific Chemicals

Bismuth hydroxide, tech., Thermo Scientific Chemicals

Catalog number: 040397.30
250 g, Each
Thermo Scientific Chemicals

Bismuth hydroxide, tech., Thermo Scientific Chemicals

Catalog number: 040397.30
250 g, Each
Quantity
Catalog number: 040397.30
also known as 040397-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10361-43-0
IUPAC Name
bismuth(3+) trihydroxide
Molecular Formula
BiH3O3
InChI Key
TZSXPYWRDWEXHG-UHFFFAOYSA-K
SMILES
[OH-].[OH-].[OH-].[Bi+3]
Assay (unspecified)
81.7-86.6% Bi

Description

It is used for the hydrolysis of ribonucleic acid and as absorbent for rutin and quercetin. It is a component used in milk of bismuth is used in gastrointestinal disorders as a protective agent. It is used for the removal of nitrate and other anions from water by yellow bismuth hydroxide. A bismuth oxide nanoplate is useful for carbon dioxide gas sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text