Thermo Scientific Chemicals

Bismuth citrate, 94%, Thermo Scientific Chemicals

Catalog number: 040398.A1
1 kg, Each
Thermo Scientific Chemicals

Bismuth citrate, 94%, Thermo Scientific Chemicals

Catalog number: 040398.A1
1 kg, Each
Quantity
Catalog number: 040398.A1
also known as 040398-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
813-93-4
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥48.3% Bismuth (equates to ≥92.0% purity)
Assay from Suppliers CofA
Appearance in solution: clear colorless
Assay from Suppliers CofA
Arsenic: Report value

Description

Bismuth citrate is used as a progressive hair dye in cosmetics. It is used as antimicrobial agents for syphills, colitis, wound infection, quartan malaria, dyspepsia and peptic ulcers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text