Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Thermo Scientific Chemicals, Premion™
Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Thermo Scientific Chemicals, Premion™
Thermo Scientific Chemicals

Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Thermo Scientific Chemicals, Premion™

Have Questions?
Quantity:
5 g
Catalog number 040401.06
also known as 040401-06
Price (USD)/ Each
755.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
755.00
Add to cart
Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Thermo Scientific Chemicals, Premion™
Catalog number040401.06
Price (USD)/ Each
755.00
-
Add to cart
Chemical Identifiers
CAS7790-46-7
IUPAC Nameplatinum(4+) tetraiodide
Molecular FormulaI4Pt
InChI KeyRNJPWBVOCUGBGY-UHFFFAOYSA-J
SMILES[I-].[I-].[I-].[I-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 27.3% min
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only