Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals
Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
0.25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040402.03
also known as 040402-03
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
Add to cart
Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min, Thermo Scientific Chemicals
Catalog number040402.03
Price (USD)/ Each
266.65
Online exclusive
296.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-15-4
IUPAC Namedioxoplatinum
Molecular FormulaO2Pt
InChI KeyYKIOKAURTKXMSB-UHFFFAOYSA-N
SMILESO=[Pt]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (special)Pt 84.4-85.6%
Appearance (Color)Brown
Total Metal Impurities≤0.05%
Platinum(IV) oxide is used as catalyst for hydrogenation reactions. It is applied in the hydrogenation of alkenes to alkanes, nitro compounds to amines and ketones to alcohols. It is also involved in the hydrogenolysis of cyclopropyl group in to an isopropyl group and in selective oxidation of primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only