Zinc bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Zinc bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc bromide, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Catalog number 040413.A1
also known as 040413-A1
Price (USD)/ Each
738.00
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
738.00
Add to cart
Zinc bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number040413.A1
Price (USD)/ Each
738.00
-
Add to cart
Chemical Identifiers
CAS7699-45-8
IUPAC Namezinc(2+) dibromide
Molecular FormulaBr2Zn
InChI KeyVNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES[Zn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
In organic chemistry as a Lewis acid. It is the electrolyte in the zinc bromide battery. Zinc bromide solutions can be used as a transparent shield against radiation. In oil and natural gas wells, solutions containing zinc bromide are used to displace drilling mud.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only