Aluminum slug, 1.98mm (0.078in) dia x 8.0mm (0.315in) length, 99.99% (metals basis), Thermo Scientific Chemicals
Aluminum slug, 1.98mm (0.078in) dia x 8.0mm (0.315in) length, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum slug, 1.98mm (0.078in) dia x 8.0mm (0.315in) length, 99.99% (metals basis), Thermo Scientific Chemicals

Evaporation or metallizing source material
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 040417.30
also known as 040417-30
Price (USD)/ Each
636.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
636.00
Add to cart
Aluminum slug, 1.98mm (0.078in) dia x 8.0mm (0.315in) length, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number040417.30
Price (USD)/ Each
636.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Trace Metalsless than 0.01% trace metals
Evaporation or metallizing source material

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only