Thermo Scientific Chemicals

Iridium(IV) iodide, Premion™, 99.95% (metals basis), Ir 27.0% min, Thermo Scientific Chemicals

Catalog number: 040424.03
1 g, Each
Thermo Scientific Chemicals

Iridium(IV) iodide, Premion™, 99.95% (metals basis), Ir 27.0% min, Thermo Scientific Chemicals

Catalog number: 040424.03
1 g, Each
Quantity
Catalog number: 040424.03
also known as 040424-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7790-45-6
Molecular Formula
I4Ir
InChI Key
UXDVYGYWEFPPTQ-UHFFFAOYSA-N
SMILES
[I].[I].[I].[I].[Ir]
Molecular Weight (g/mol)
699.84
Total Metal Impurities
0.05% max.

Description

Methanol carbonylation to acetic acid is catalysed with high rates at low water concentrations using an iridium/iodide based catalyst. Iridium(IV)-iodide mixtures participates in the Investigations in platinum metal group electrochemistry. Rhodium- and iridium-iodide catalysts also promote the carbonylation of higher alcohols to linear or branched chain carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text