Thermo Scientific Chemicals

Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Thermo Scientific Chemicals, Premion™

Catalog number: 040428.03
1 g, Each
Thermo Scientific Chemicals

Sodium tetrabromoaurate(III) hydrate, 99.99% (Metals basis), Thermo Scientific Chemicals, Premion™

Catalog number: 040428.03
1 g, Each
Quantity
Catalog number: 040428.03
also known as 040428-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10378-49-1
IUPAC Name
sodium tetrabromogolduide
Molecular Formula
AuBr4Na
InChI Key
PMPPMRNISYJZFW-UHFFFAOYSA-J
SMILES
[Na+].Br[Au-](Br)(Br)Br
Total Metal Impurities
0.01% max.

Description

Sodium tetrabromoaurate(III) hydrate participates in the fabricated of core/alloy nanoparticles via the layer-by-layer hydrothermal route ans its growth characteristics and optical properties study.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text