Nickel Chromium wire, 1.0mm (0.04in) dia, 99% (metals basis), Thermo Scientific Chemicals
Nickel Chromium wire, 1.0mm (0.04in) dia, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel Chromium wire, 1.0mm (0.04in) dia, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
25 m
500 m
Catalog number 040438.H4
also known as 040438-H4
Price (USD)/ Each
177.00
-
Add to cart
Quantity:
100 m
Request bulk or custom format
Price (USD)/ Each
177.00
Add to cart
Nickel Chromium wire, 1.0mm (0.04in) dia, 99% (metals basis), Thermo Scientific Chemicals
Catalog number040438.H4
Price (USD)/ Each
177.00
-
Add to cart
Chemical Identifiers
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Comment1.0 mm diameter
CommentNi 80% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only