Thermo Scientific Chemicals

Buffer solution, pH 2.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Catalog number: 040447.K7
4 L, Each
Thermo Scientific Chemicals

Buffer solution, pH 2.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable, Thermo Scientific Chemicals

Catalog number: 040447.K7
4 L, Each
Quantity
Catalog number: 040447.K7
also known as 040447-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7, 7647-01-0, 7732-18-5

Description

Buffer Solution pH 2 used as a means of keeping pH at a nearly constant value in a different chemical applications. Glycine Buffer Solution pH 2 used for eluting antibodies in various antibody purification procedures i.e. protein A-agarose, protein G-agarose, affinity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text