Thermo Scientific Chemicals

Cerium(III) 2-ethylhexanoate, 49% in 2-ethylhexanoic acid, Ce 12%, Thermo Scientific Chemicals

Catalog number: 040451.22
100 g, Each
Thermo Scientific Chemicals

Cerium(III) 2-ethylhexanoate, 49% in 2-ethylhexanoic acid, Ce 12%, Thermo Scientific Chemicals

Catalog number: 040451.22
100 g, Each
Quantity
Catalog number: 040451.22
also known as 040451-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56797-01-4
IUPAC Name
cerium(3+) tris(2-ethylhexanoate)
Molecular Formula
C24H45CeO6
InChI Key
GGVUYAXGAOIFIC-UHFFFAOYNA-K
SMILES
[Ce+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Appearance (Color)
Clear colorless to yellow
Assay (special)
Cerium, Ce: 11.8-12.2% (theory for 49% solution: 12.05%)
Form
Viscous liquid

Description

Used as catalyst, paint dryer, plastic Additive. As the the precursor in preparation of inorganic nanoparticles in oil-in-water microemulsions. The Toluene-based precursor solution was composed by mixing gadolinium (III) 2-ethylhexanoate and cerium (III) 2-ethylhexanoate to achieve the right stoichiometry. For the ceria/zirconia precursor, cerium(III) 2-ethylhexanoate and zirconium(IV) 2-ethylhexanoate were mixed to result in a metal molar ratio Ce/Zr of 1:1 and diluted with xylene to a total metal. Thin films of CeO 2 were prepared by a photochemical method using thin amorphous films of cerium(III) 2-ethylhexanoate as the precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text