Thermo Scientific Chemicals

Rhenium(VII) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040468.18
50 g, Each
Thermo Scientific Chemicals

Rhenium(VII) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 040468.18
50 g, Each
Quantity
Catalog number: 040468.18
also known as 040468-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-68-7
IUPAC Name
trioxo[(trioxorhenio)oxy]rhenium
Molecular Formula
O7Re2
InChI Key
NBGOSNNAAHDRLK-UHFFFAOYSA-N
SMILES
O=[Re](=O)(=O)O[Re](=O)(=O)=O
Total Metal Impurities
≤0.1% (1000 ppm)
Form
Crystalline
Appearance (Color)
Yellow to black

Description

As a catalyst for oxidation/reduction reactionsRhenium(VII) oxide acts as a precursor to methylrhenium trioxide, organorhenium oxides which catalyze olefin oxidation, metathesis and other transformations. It is used as a reagent for dehydrations and rearrangements of allylic alcohols, oximes and amides, oxidative cyclizations, alkene metathesis and formation of bisperoxyacetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text