Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min, Thermo Scientific Chemicals
Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min, Thermo Scientific Chemicals

Ceramic glazes and enamel fluxes
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 040476.14
also known as 040476-14
Price (USD)/ Each
37.65
Online exclusive
41.50 
Save 3.85 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.50 
Save 3.85 (9%)
Add to cart
Bismuth hydroxide nitrate oxide, Bi{2}O{3} 79% min, Thermo Scientific Chemicals
Catalog number040476.14
Price (USD)/ Each
37.65
Online exclusive
41.50 
Save 3.85 (9%)
-
Add to cart
Chemical Identifiers
CAS1304-85-4
IUPAC Namebis(nitrooxy)bismuthanyl nitrate
Molecular FormulaBiN3O9
InChI KeyPPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES[O-][N+](=O)O[Bi](O[N+]([O-])=O)O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofABi₂O₃: ≥79.0%
It is used pharmaceutically in wound dressing, ointment component and as an antiacid. In cosmetics it is used as a cosmetic white pigment. Used in Ceramic glazes and enamel fluxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only