Thermo Scientific Chemicals

tert-Butyl methyl ether, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 040477.M6
4 x 4 L, Each
Thermo Scientific Chemicals

tert-Butyl methyl ether, HPLC Grade, 99+%, Thermo Scientific Chemicals

Catalog number: 040477.M6
4 x 4 L, Each
Quantity
Catalog number: 040477.M6
also known as 040477-M6
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1634-04-4
IUPAC Name
2-methoxy-2-methylpropane
Molecular Formula
C5H12O
InChI Key
BZLVMXJERCGZMT-UHFFFAOYSA-N
SMILES
COC(C)(C)C
Form
Liquid
Color (APHA)
≤10
Comment
Peroxide (as H2O2): ≤1ppm
Residual water
≤0.05%
Optical Absorbance at
210nm: <0.50

Description

Used as a Chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text