Niobium powder, APS 1-5 micron, 99.8% (metals basis), Thermo Scientific Chemicals
Niobium powder, APS 1-5 micron, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium powder, APS 1-5 micron, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040478.30
also known as 040478-30
Price (USD)/ Each
388.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
457.00 
Save 68.35 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
388.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
457.00 
Save 68.35 (15%)
Add to cart
Niobium powder, APS 1-5 micron, 99.8% (metals basis), Thermo Scientific Chemicals
Catalog number040478.30
Price (USD)/ Each
388.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
457.00 
Save 68.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size1-5 microns average
FormPowder
Total Metal Impurities0.2% max.
Used in high temperature and super alloys of Zn and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only