Thermo Scientific Chemicals
ICP-MS Stock Standard solution A for 200.8, Rev. 4.4, Specpure™, Thermo Scientific Chemicals
Catalog number: 040485.AE
100 mL, Each
Thermo Scientific Chemicals
ICP-MS Stock Standard solution A for 200.8, Rev. 4.4, Specpure™, Thermo Scientific Chemicals
Catalog number: 040485.AE
100 mL, Each
Quantity
Catalog number: 040485.AE
also known as 040485-AE
Price (USD)
Have Questions?
Concentration
Pb 10?g/mL ?2%
Concentration
V 10?g/mL ?2%
Concentration
Ni 10?g/mL ?2%
Concentration
Se 10?g/mL ?2%
Concentration
U 10?g/mL ?2%