Iron foil, 1.0mm (0.04in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Iron foil, 1.0mm (0.04in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron foil, 1.0mm (0.04in) thick, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 040496.FF
also known as 040496-FF
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
Add to cart
Iron foil, 1.0mm (0.04in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number040496.FF
Price (USD)/ Each
157.65
Online exclusive
175.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.99% (metals basis)
FormFoil, 1.0mm (0.04in) thick.
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only