Thermo Scientific Chemicals

Tungsten carbide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040502.30
250 g, Each
Thermo Scientific Chemicals

Tungsten carbide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040502.30
250 g, Each
Quantity
Catalog number: 040502.30
also known as 040502-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12070-12-1
IUPAC Name
methanidylidynetungstenylium
Molecular Formula
CW
InChI Key
UONOETXJSWQNOL-UHFFFAOYSA-N
SMILES
[C-]#[W+]
Total Metal Impurities
1% max.

Description

Sintered tungsten carbide cutting tools are very abrasion resistant. Tungsten carbide is often used in armor-piercing ammunition. Tungsten carbide is also an effective neutron reflector. Hard carbides, especially tungsten carbide, are used by athletes for balance. Tungsten carbide is also used for making surgical instruments

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text