Boron carbide, Thermo Scientific Chemicals
Boron carbide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron carbide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040504.30
also known as 040504-30
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
Add to cart
Boron carbide, Thermo Scientific Chemicals
Catalog number040504.30
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS12069-32-8
IUPAC Name2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
Molecular FormulaCB4
InChI KeyINAHAJYZKVIDIZ-UHFFFAOYSA-N
SMILESB12B3B4B1C234
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofAReport: % Boron Carbide (B4C)
Particle Size<10 micron (APS)
Appearance (Color)Black
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only