Thermo Scientific Chemicals

Ammonium sulfide, 40-44% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 040509.AE
100 mL, Each
Thermo Scientific Chemicals

Ammonium sulfide, 40-44% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: 040509.AE
100 mL, Each
Quantity
Catalog number: 040509.AE
also known as 040509-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12135-76-1
IUPAC Name
diamine sulfane
Molecular Formula
H8N2S
InChI Key
UXXGWUQNRMPNKH-UHFFFAOYSA-N
SMILES
N.N.S
Concentration (weight/weight)
40-44%

Description

Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth.It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text