Bismuth Lead Tin ingot (Rose's metal), Thermo Scientific Chemicals
Bismuth Lead Tin ingot (Rose's metal), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth Lead Tin ingot (Rose's metal), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040516.36
also known as 040516-36
Price (USD)/ Each
612.65
Online exclusive
681.00 
Save 68.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
612.65
Online exclusive
681.00 
Save 68.35 (10%)
Add to cart
Bismuth Lead Tin ingot (Rose's metal), Thermo Scientific Chemicals
Catalog number040516.36
Price (USD)/ Each
612.65
Online exclusive
681.00 
Save 68.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialBismuth Lead Tin ingot (Rose's metal)
CAS56001-37-7
Melting Point95°C to 96°C
Health Hazard 1H302+H332-H360Df-H373
Health Hazard 2GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
View more
Rose's metal was used as the droplet fluid, because it has a low melting point (e460 K). Rose's metal is used as a sample in computer-controlled diffractometer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only