Thermo Scientific Chemicals

Chromyl chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040517.18
50 g, Each
Thermo Scientific Chemicals

Chromyl chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 040517.18
50 g, Each
Quantity
Catalog number: 040517.18
also known as 040517-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14977-61-8
IUPAC Name
λ²-chromium(2+) dihydrate dichloride
Molecular Formula
Cl2CrH4O2
InChI Key
HOCPFYCTFLRIAK-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Cr++]
Total Metal Impurities
0.01% max.

Description

Highly spin-polarized chromium dioxide (CrO2) thin films were deposited on 100 TiO2 substrates by chemical vapor deposition using chromyl chloride as a precursor. Chromyl chloride (Cr02C12) reacts with cyclohexane solvent at 75 OC to give a dark precipitate along with chlorocyclohexane and a small amount of cyclohexene (in 10.0 and ca. 0.3% yields based on chromium). Chromyl chloride, CrO2C12, oxidizes cyclooctane, isobutane, and toluene under mild conditions (25-60 C). The reactions give chlorinated products (chlorocyclooctane, tert- butyl chloride, and benzyl chloride) and a dark chromium-containing precipitate. Used for Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins. Chromyl chloride in carbon tetrachloride solution reacts in the cold with cyclohexene, cyclopentene, and 1-hexene to give the various isomeric chlorohydrins as the major products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text