Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals
Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number 040522.06
also known as 040522-06
Price (USD)/ Each
537.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
537.00
Add to cart
Cyclopentadienylmolybdenum(V) tetrachloride, 95%, Thermo Scientific Chemicals
Catalog number040522.06
Price (USD)/ Each
537.00
-
Add to cart
Specifications
Chemical Name or MaterialCyclopentadienylmolybdenum(V) tetrachloride
CAS62927-99-5
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only