Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 70,000, 30% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 040529.36
500 g, Each
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 70,000, 30% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 040529.36
500 g, Each
Quantity
Catalog number: 040529.36
also known as 040529-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-98-6
Molecular Formula
((C2H5N)zC2H4N)y(C2H5N)x
SMILES
*-CCNCCN(-*)CCN-*
MDL Number
MFCD00084427
CAS
9002-98-6
Appearance (Color)
Colorless to Yellow
Assay from Supplier's CofA
29.5-30.5%
Viscosity
400 - 900 cps (25°C)
Appearance (Color)
Colorless to Yellow
Form
Liquid

Description

Acts as a protein precipitant used to purify proteins. It is used as chelating agent and as a scavenger for aldehydes and oxides. Also used in detergents, paper industry, dyes, printing inks and in water treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text