Sulfur in Kerosene standard solution, Specpure™ blank (0.000%), Thermo Scientific Chemicals
Sulfur in Kerosene standard solution, Specpure™ blank (0.000%), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur in Kerosene standard solution, Specpure™ blank (0.000%), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 040536.AE
also known as 040536-AE
Price (USD)/ Each
91.60
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
91.60
Add to cart
Sulfur in Kerosene standard solution, Specpure™ blank (0.000%), Thermo Scientific Chemicals
Catalog number040536.AE
Price (USD)/ Each
91.60
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialSulfur in Kerosene standard solution
Name NoteBlank (0.000%)
Health Hazard 1H227-H304
Quantity100 mL
Health Hazard 2GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only