Thermo Scientific Chemicals

Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040568.ND
25 g, Each
Thermo Scientific Chemicals

Neodymium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040568.ND
25 g, Each
Quantity
Catalog number: 040568.ND
also known as 040568-ND
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17522-69-9
IUPAC Name
neodymium(3+) triperchlorate
Molecular Formula
Cl3NdO12
InChI Key
HBKMAYJLXKBOER-UHFFFAOYSA-K
SMILES
[Nd+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Form
50% Aqueous Solution
Assay from Suppliers CofA
Neodymium content: 12.6-13.6%
Chloride content
≤0.01%
Insoluble material
≤0.01%

Description

Neodymium(III) perchlorate used in the preparation of complexes of phosphine oxides. Two three-dimensional frameworks composed of doubly interpenetrating networks of the same topology but different stereoisomeric structures have been constructed from self-assembly of the chiral building block, Δ- or Λ-[Nd(ntb)2]3+.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text