Thermo Scientific Chemicals

Terbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040573.NC
10 g, Each
Thermo Scientific Chemicals

Terbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040573.NC
10 g, Each
Quantity
Catalog number: 040573.NC
also known as 040573-NC
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14014-09-6
IUPAC Name
terbium(3+) hexahydrate triperchlorate
Molecular Formula
Cl3H12O18Tb
InChI Key
NAPXFYQZNZUQDE-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Tb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Concentration (weight/weight)
50% min

Description

In observation of slow magnetic relaxation in triple-stranded lanthanide helicates Terbium(III) perchlorate is substituted for dysprosium(III) perchlorate hexahydrate. By reacting 3-formylchromone and hydrated terbium (III) perchlorate in isopropanol, 3-hydroxymethylchromone [icm], bis(3-chromonyl)methanol [dcm] and the novel complex Tb(imc)3(ClO4)3.5H2O are obtained.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text