Thermo Scientific Chemicals

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040576.NA
∼2 g, Each
Thermo Scientific Chemicals

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific Chemicals

Catalog number: 040576.NA
∼2 g, Each
Quantity
Catalog number: 040576.NA
also known as 040576-NA
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14692-15-0
IUPAC Name
erbium(3+) hexahydrate triperchlorate
Molecular Formula
Cl3ErH12O18
InChI Key
JZRNHNTYRRYOCR-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Er+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Concentration (weight/weight)
50% min

Description

Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text