Cerium(IV) oxide, 96% (REO), Thermo Scientific Chemicals
Cerium(IV) oxide, 96% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(IV) oxide, 96% (REO), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 040581.A1
also known as 040581-A1
Price (USD)/ Each
469.65
Online exclusive
522.00 
Save 52.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
469.65
Online exclusive
522.00 
Save 52.35 (10%)
Add to cart
Cerium(IV) oxide, 96% (REO), Thermo Scientific Chemicals
Catalog number040581.A1
Price (USD)/ Each
469.65
Online exclusive
522.00 
Save 52.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1306-38-3
IUPAC Namedioxocerium
Molecular FormulaCeO2
InChI KeyCETPSERCERDGAM-UHFFFAOYSA-N
SMILESO=[Ce]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities4% max.
Cerium(IV) oxide is used in polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions
RUO – Research Use Only